Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTTCCTTCCTTATTCTACCGGCA[C/T]AAAGGGATCTTCAACTCGCATCTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606283 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SORCS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SORCS1 - sortilin related VPS10 domain containing receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013031.2 | 4634 | UTR 3 | NP_001013049.1 | |||
NM_001206569.1 | 4634 | Intron | NP_001193498.1 | |||
NM_001206570.1 | 4634 | UTR 3 | NP_001193499.1 | |||
NM_001206571.1 | 4634 | Intron | NP_001193500.1 | |||
NM_001206572.1 | 4634 | UTR 3 | NP_001193501.1 | |||
NM_052918.4 | 4634 | UTR 3 | NP_443150.3 | |||
XM_011539199.2 | 4634 | Intron | XP_011537501.1 | |||
XM_011539201.2 | 4634 | Intron | XP_011537503.1 | |||
XM_017015614.1 | 4634 | Intron | XP_016871103.1 | |||
XM_017015615.1 | 4634 | Intron | XP_016871104.1 | |||
XM_017015616.1 | 4634 | Intron | XP_016871105.1 | |||
XM_017015617.1 | 4634 | Intron | XP_016871106.1 | |||
XM_017015618.1 | 4634 | Intron | XP_016871107.1 |