Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAAGTGGTCTCCGCCTTATCTGG[A/G]GTGCGAGAACGGGAGTTACAAGCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614573 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR158 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPR158 - G protein-coupled receptor 158 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020752.2 | 872 | Missense Mutation | GAG,GGG | E,G 271 | NP_065803.2 | |
XM_017016452.1 | 872 | Intron | XP_016871941.1 |
GPR158-AS1 - GPR158 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |