Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTCCAAACCAAAACTAATTACTC[A/G]GTTGGAGCAAGGGGCAGAGCCCTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF485 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF485 - zinc finger protein 485 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318140.1 | 1715 | Missense Mutation | CAG,CGG | Q,R 62 | NP_001305069.1 | |
NM_001318141.1 | 1715 | Missense Mutation | CAG,CGG | Q,R 62 | NP_001305070.1 | |
NM_001318142.1 | 1715 | UTR 5 | NP_001305071.1 | |||
NM_001318143.1 | 1715 | Intron | NP_001305072.1 | |||
NM_145312.3 | 1715 | Missense Mutation | CAG,CGG | Q,R 62 | NP_660355.2 | |
XM_011539498.2 | 1715 | UTR 5 | XP_011537800.1 |