Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTGTCTGGCCCAGCAGCAGCCTC[G/T]GCAGGGAGCATGGTAGTGGACACGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607506 | ||||||||||||||||||||
Literature Links: |
ADAMTS14 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADAMTS14 - ADAM metallopeptidase with thrombospondin type 1 motif 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080722.4 | 477 | Silent Mutation | TCG,TCT | S,S 64 | NP_542453.2 | |
NM_139155.3 | 477 | Silent Mutation | TCG,TCT | S,S 64 | NP_631894.2 | |
XM_011539300.2 | 477 | Silent Mutation | TCG,TCT | S,S 64 | XP_011537602.1 | |
XM_011539301.2 | 477 | Intron | XP_011537603.1 | |||
XM_011539302.2 | 477 | Intron | XP_011537604.1 | |||
XM_011539303.2 | 477 | Silent Mutation | TCG,TCT | S,S 64 | XP_011537605.1 | |
XM_011539305.2 | 477 | Silent Mutation | TCG,TCT | S,S 64 | XP_011537607.1 | |
XM_011539306.2 | 477 | Silent Mutation | TCG,TCT | S,S 64 | XP_011537608.1 | |
XM_011539307.2 | 477 | Silent Mutation | TCG,TCT | S,S 64 | XP_011537609.1 | |
XM_011539308.2 | 477 | Silent Mutation | TCG,TCT | S,S 64 | XP_011537610.1 | |
XM_011539309.1 | 477 | Intron | XP_011537611.1 |