Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTATGACCTGAGTTTCCTCAATAGG[A/G]TGGAGTTTTCTTAGAACCGGCTGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601403 | ||||||||||||||||||||
Literature Links: |
DOCK1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DOCK1 - dedicator of cytokinesis 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290223.1 | 5444 | Intron | NP_001277152.1 | |||
NM_001380.4 | 5444 | Intron | NP_001371.1 | |||
XM_011539422.2 | 5444 | Intron | XP_011537724.1 | |||
XM_011539424.1 | 5444 | Intron | XP_011537726.1 | |||
XM_017015813.1 | 5444 | Intron | XP_016871302.1 | |||
XM_017015814.1 | 5444 | Intron | XP_016871303.1 | |||
XM_017015815.1 | 5444 | Intron | XP_016871304.1 | |||
XM_017015816.1 | 5444 | Intron | XP_016871305.1 | |||
XM_017015817.1 | 5444 | Intron | XP_016871306.1 |
FAM196A - family with sequence similarity 196 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039762.2 | 5444 | Silent Mutation | CAC,CAT | H,H 439 | NP_001034851.1 | |
XM_005252694.3 | 5444 | Silent Mutation | CAC,CAT | H,H 439 | XP_005252751.1 | |
XM_017016537.1 | 5444 | Silent Mutation | CAC,CAT | H,H 439 | XP_016872026.1 | |
XM_017016538.1 | 5444 | Silent Mutation | CAC,CAT | H,H 439 | XP_016872027.1 | |
XM_017016539.1 | 5444 | Silent Mutation | CAC,CAT | H,H 439 | XP_016872028.1 | |
XM_017016540.1 | 5444 | Silent Mutation | CAC,CAT | H,H 439 | XP_016872029.1 | |
XM_017016541.1 | 5444 | Silent Mutation | CAC,CAT | H,H 439 | XP_016872030.1 | |
XM_017016542.1 | 5444 | Silent Mutation | CAC,CAT | H,H 439 | XP_016872031.1 | |
XM_017016543.1 | 5444 | Intron | XP_016872032.1 |