Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAACAGGTGGAGGAGGATGGAAG[A/G]TCTCCCTGGTCGGAGGCCTAAAATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607942 MIM: 616872 | ||||||||||||||||||||
Literature Links: |
PIK3AP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PIK3AP1 - phosphoinositide-3-kinase adaptor protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152309.2 | 2053 | Missense Mutation | ACC,ATC | T,I 793 | NP_689522.2 | |
XM_005269498.1 | 2053 | Missense Mutation | ACC,ATC | T,I 615 | XP_005269555.1 | |
XM_005269499.1 | 2053 | Missense Mutation | ACC,ATC | T,I 615 | XP_005269556.1 | |
XM_011539248.1 | 2053 | Silent Mutation | GAC,GAT | D,D 753 | XP_011537550.1 | |
XM_011539249.1 | 2053 | Missense Mutation | ACC,ATC | T,I 741 | XP_011537551.1 |
TM9SF3 - transmembrane 9 superfamily member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |