Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGGAGCGCAGTCGTGCACTGGGC[G/T]TGGCGCTGGCCTTCATTAGCTTCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 118490 MIM: 600336 | ||||||||||||||||||||
Literature Links: |
CHAT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHAT - choline O-acetyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142929.1 | 1086 | Intron | NP_001136401.1 | |||
NM_001142933.1 | 1086 | Intron | NP_001136405.1 | |||
NM_001142934.1 | 1086 | Intron | NP_001136406.1 | |||
NM_020549.4 | 1086 | Intron | NP_065574.3 | |||
NM_020984.3 | 1086 | Intron | NP_066264.3 | |||
NM_020985.3 | 1086 | Intron | NP_066265.3 | |||
NM_020986.3 | 1086 | Intron | NP_066266.3 |
SLC18A3 - solute carrier family 18 member A3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003055.2 | 1086 | Missense Mutation | GTG,TTG | V,L 216 | NP_003046.2 |