Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTCCTGCAGATGAAGCGCAGGAA[A/G]TACGGCTTCATCTACAAGACGCATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602239 MIM: 608428 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP26A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYP26A1 - cytochrome P450 family 26 subfamily A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000783.3 | 267 | Silent Mutation | AAA,AAG | K,K 74 | NP_000774.2 | |
NM_057157.2 | 267 | Silent Mutation | AAA,AAG | K,K 5 | NP_476498.1 |
CYP26C1 - cytochrome P450 family 26 subfamily C member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |