Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGATTTGTTAAGGCCATAGCTGCG[C/T]TGATCTCCTTCCAATACGTGTCATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601332 | ||||||||||||||||||||
Literature Links: |
MKX PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MKX - mohawk homeobox | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242702.1 | 1151 | Missense Mutation | AAC,AGC | N,S 309 | NP_001229631.1 | |
NM_173576.2 | 1151 | Missense Mutation | AAC,AGC | N,S 309 | NP_775847.2 | |
XM_017016105.1 | 1151 | Missense Mutation | AAC,AGC | N,S 309 | XP_016871594.1 | |
XM_017016106.1 | 1151 | Missense Mutation | AAC,AGC | N,S 309 | XP_016871595.1 | |
XM_017016107.1 | 1151 | Intron | XP_016871596.1 | |||
XM_017016108.1 | 1151 | UTR 3 | XP_016871597.1 |