Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGCGAGGAGCGCTTCCGCGTGGA[C/T]AGGAAGAAACTTGAGGCCATGTTAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614295 | ||||||||||||||||||||
Literature Links: |
BICC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BICC1 - BicC family RNA binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080512.2 | 162 | Silent Mutation | GAC,GAT | D,D 54 | NP_001073981.1 | |
XM_005270169.4 | 162 | Intron | XP_005270226.1 | |||
XM_011540185.2 | 162 | Silent Mutation | GAC,GAT | D,D 54 | XP_011538487.1 | |
XM_011540188.2 | 162 | Intron | XP_011538490.2 | |||
XM_011540189.2 | 162 | Intron | XP_011538491.1 | |||
XM_011540190.2 | 162 | Intron | XP_011538492.1 | |||
XM_011540191.2 | 162 | Intron | XP_011538493.1 | |||
XM_017016677.1 | 162 | Intron | XP_016872166.1 | |||
XM_017016678.1 | 162 | Intron | XP_016872167.1 |