Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACAACAGCTTACCCTGCAGGGTAT[A/G]CAGAAGCTGGAGGAAGGTCTGATAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609177 | ||||||||||||||||||||
Literature Links: |
ZWINT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZWINT - ZW10 interacting kinetochore protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005413.1 | 686 | Intron | NP_001005413.1 | |||
NM_007057.3 | 686 | Missense Mutation | CAT,TAT | H,Y 217 | NP_008988.2 | |
NM_032997.2 | 686 | Missense Mutation | CAT,TAT | H,Y 217 | NP_127490.1 | |
XM_017015605.1 | 686 | Intron | XP_016871094.1 |