Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAGTACCTTCCATTATCCTTTCA[A/G]TGCCAATGAAATGATAGTATTAAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608909 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARL5B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARL5B - ADP ribosylation factor like GTPase 5B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178815.3 | Intron | NP_848930.1 | ||||
XM_005252400.1 | Intron | XP_005252457.1 | ||||
XM_005252401.4 | Intron | XP_005252458.1 |
NSUN6 - NOP2/Sun RNA methyltransferase family member 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |