Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGACTTCACGTGCCCGTTTCCTT[C/T]GCAGTCTCTCAGCATTTGTGATCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611206 MIM: 609204 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFAP70 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CFAP70 - cilia and flagella associated protein 70 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DNAJC9 - DnaJ heat shock protein family (Hsp40) member C9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DNAJC9-AS1 - DNAJC9 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FAM149B1 - family with sequence similarity 149 member B1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPS16 - mitochondrial ribosomal protein S16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016065.3 | Intron | NP_057149.1 | ||||
XM_017016305.1 | Intron | XP_016871794.1 |