Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTCATGCGGATGATTTATTCCCA[A/T]TTTTCTTGAGACATGCCAAACAAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605122 | ||||||||||||||||||||
Literature Links: |
SUPV3L1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SUPV3L1 - Suv3 like RNA helicase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301683.1 | 617 | Missense Mutation | ATT,TTT | I,F 30 | NP_001288612.1 | |
NM_001323584.1 | 617 | Missense Mutation | ATT,TTT | I,F 30 | NP_001310513.1 | |
NM_001323585.1 | 617 | Missense Mutation | ATT,TTT | I,F 40 | NP_001310514.1 | |
NM_001323586.1 | 617 | Missense Mutation | ATT,TTT | I,F 40 | NP_001310515.1 | |
NM_001323587.1 | 617 | UTR 5 | NP_001310516.1 | |||
NM_001323588.1 | 617 | UTR 5 | NP_001310517.1 | |||
NM_003171.4 | 617 | Missense Mutation | ATT,TTT | I,F 161 | NP_003162.2 |