Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGCTACAGTCTTCACATATATGT[A/G]GGCCTCTTGTGCTCCCATCGTATTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 187410 | ||||||||||||||||||||
Literature Links: |
DNTT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DNTT - DNA nucleotidylexotransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OPALIN - oligodendrocytic myelin paranodal and inner loop protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040102.2 | 483 | Missense Mutation | CAC,TAC | H,Y 79 | NP_001035191.1 | |
NM_001040103.2 | 483 | Missense Mutation | CAC,TAC | H,Y 92 | NP_001035192.1 | |
NM_001284320.1 | 483 | Missense Mutation | CAC,TAC | H,Y 91 | NP_001271249.1 | |
NM_001284321.1 | 483 | Missense Mutation | CAC,TAC | H,Y 90 | NP_001271250.1 | |
NM_001284322.1 | 483 | Missense Mutation | CAC,TAC | H,Y 92 | NP_001271251.1 | |
NM_001284323.1 | 483 | Missense Mutation | CAC,TAC | H,Y 92 | NP_001271252.1 | |
NM_001284324.1 | 483 | Missense Mutation | CAC,TAC | H,Y 92 | NP_001271253.1 | |
NM_001284326.1 | 483 | Missense Mutation | CAC,TAC | H,Y 40 | NP_001271255.1 | |
NM_001284327.1 | 483 | Missense Mutation | CAC,TAC | H,Y 40 | NP_001271256.1 | |
NM_033207.4 | 483 | Missense Mutation | CAC,TAC | H,Y 102 | NP_149984.1 |