Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGATATCCTGTCTTGGAACTCTGC[C/T]GTGGGTACAATGGGAATAGTTCCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138130 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLUD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GLUD1 - glutamate dehydrogenase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318900.1 | 2101 | Silent Mutation | ACA,ACG | T,T 357 | NP_001305829.1 | |
NM_001318901.1 | 2101 | Silent Mutation | ACA,ACG | T,T 323 | NP_001305830.1 | |
NM_001318902.1 | 2101 | Silent Mutation | ACA,ACG | T,T 323 | NP_001305831.1 | |
NM_001318904.1 | 2101 | Silent Mutation | ACA,ACG | T,T 323 | NP_001305833.1 | |
NM_001318905.1 | 2101 | Silent Mutation | ACA,ACG | T,T 323 | NP_001305834.1 | |
NM_001318906.1 | 2101 | Silent Mutation | ACA,ACG | T,T 323 | NP_001305835.1 | |
NM_005271.4 | 2101 | Silent Mutation | ACA,ACG | T,T 490 | NP_005262.1 |