Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCAAACAGCACTATTCTGGATGC[C/T]AGCGTTTCTCTAACAGCCTAGCAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615564 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM45A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM45A - family with sequence similarity 45 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SFXN4 - sideroflexin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_213649.1 | 923 | Silent Mutation | CTA,CTG | L,L 250 | NP_998814.1 | |
XM_005269525.4 | 923 | Silent Mutation | CTA,CTG | L,L 241 | XP_005269582.1 | |
XM_005269526.2 | 923 | Silent Mutation | CTA,CTG | L,L 134 | XP_005269583.1 | |
XM_005269527.1 | 923 | Silent Mutation | CTA,CTG | L,L 134 | XP_005269584.1 | |
XM_011539282.2 | 923 | Silent Mutation | CTA,CTG | L,L 134 | XP_011537584.1 |