Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCGGTAGCCAGAATGCAGGGGCA[C/T]GGGGCGGGCGAAGTAGAGCTGGTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
PYROXD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PYROXD2 - pyridine nucleotide-disulphide oxidoreductase domain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032709.2 | 1653 | Missense Mutation | NP_116098.2 | |||
XM_011540293.2 | 1653 | Missense Mutation | XP_011538595.1 | |||
XM_011540301.2 | 1653 | Missense Mutation | XP_011538603.1 | |||
XM_017016835.1 | 1653 | Missense Mutation | XP_016872324.1 | |||
XM_017016836.1 | 1653 | Intron | XP_016872325.1 | |||
XM_017016837.1 | 1653 | Intron | XP_016872326.1 | |||
XM_017016838.1 | 1653 | Missense Mutation | XP_016872327.1 | |||
XM_017016839.1 | 1653 | Missense Mutation | XP_016872328.1 | |||
XM_017016840.1 | 1653 | Missense Mutation | XP_016872329.1 | |||
XM_017016841.1 | 1653 | Missense Mutation | XP_016872330.1 |