Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAAAGGCAAAAAAGAAAGAGGAG[C/G]CATCTCAAAATGACATTTCTCCTAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606357 MIM: 610373 | ||||||||||||||||||||
Literature Links: |
DDX21 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DDX21 - DEAD-box helicase 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256910.1 | 469 | Missense Mutation | CCA,GCA | P,A 15 | NP_001243839.1 | |
NM_004728.3 | 469 | Missense Mutation | CCA,GCA | P,A 83 | NP_004719.2 | |
XM_011540336.1 | 469 | Missense Mutation | CCA,GCA | P,A 83 | XP_011538638.1 | |
XM_017016910.1 | 469 | Missense Mutation | CCA,GCA | P,A 15 | XP_016872399.1 |
DDX50 - DEAD-box helicase 50 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |