Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGCTGTGTGTTTCCACAGCGCCA[A/G]CAGCAACGCGGACCACGTAGGTGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611144 | ||||||||||||||||||||
Literature Links: |
FAM175B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM175B - family with sequence similarity 175 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032182.3 | 72 | Missense Mutation | AAC,AGC | N,S 19 | NP_115558.3 | |
XM_011539554.1 | 72 | UTR 5 | XP_011537856.1 | |||
XM_017015986.1 | 72 | UTR 5 | XP_016871475.1 |
METTL10 - methyltransferase like 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |