Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGAGTCTTTAAGAAGTCCTGGAAC[A/T]CTTCCTAACCACCCTAATTTCAGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605796 | ||||||||||||||||||||
Literature Links: |
TDRD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TDRD1 - tudor domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198795.1 | 305 | Silent Mutation | ACA,ACT | T,T 46 | NP_942090.1 | |
XM_005269978.2 | 305 | Silent Mutation | ACA,ACT | T,T 46 | XP_005270035.1 | |
XM_005269979.2 | 305 | Silent Mutation | ACA,ACT | T,T 46 | XP_005270036.1 | |
XM_011539959.2 | 305 | Silent Mutation | ACA,ACT | T,T 46 | XP_011538261.1 | |
XM_011539960.2 | 305 | Silent Mutation | ACA,ACT | T,T 46 | XP_011538262.1 | |
XM_011539961.2 | 305 | Silent Mutation | ACA,ACT | T,T 46 | XP_011538263.1 | |
XM_011539962.1 | 305 | Silent Mutation | ACA,ACT | T,T 46 | XP_011538264.1 | |
XM_011539963.1 | 305 | Silent Mutation | ACA,ACT | T,T 46 | XP_011538265.1 | |
XM_011539964.1 | 305 | Silent Mutation | ACA,ACT | T,T 46 | XP_011538266.1 | |
XM_011539966.1 | 305 | Silent Mutation | ACA,ACT | T,T 46 | XP_011538268.1 | |
XM_017016414.1 | 305 | Silent Mutation | ACA,ACT | T,T 46 | XP_016871903.1 | |
XM_017016415.1 | 305 | Silent Mutation | ACA,ACT | T,T 46 | XP_016871904.1 |