Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGGCCAGCTCCTCCCCGCCACCA[C/T]GGCCCTCCTGGGCCCACCTTCTTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606075 MIM: 610454 MIM: 612971 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C10orf2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C10orf2 - chromosome 10 open reading frame 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LZTS2 - leucine zipper tumor suppressor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318099.1 | 356 | Silent Mutation | CAC,CAT | H,H 48 | NP_001305028.1 | |
NM_001318100.1 | 356 | Silent Mutation | CAC,CAT | H,H 48 | NP_001305029.1 | |
NM_001318101.1 | 356 | Silent Mutation | CAC,CAT | H,H 48 | NP_001305030.1 | |
NM_032429.3 | 356 | Silent Mutation | CAC,CAT | H,H 48 | NP_115805.1 | |
XM_005270222.4 | 356 | Missense Mutation | CAC,CAT | H,H 48 | XP_005270279.1 | |
XM_017016780.1 | 356 | Missense Mutation | CAC,CAT | H,H 48 | XP_016872269.1 | |
XM_017016781.1 | 356 | Missense Mutation | CAC,CAT | H,H 48 | XP_016872270.1 |
PDZD7 - PDZ domain containing 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |