Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTAGACCTAAAGTCCAGATTAAAT[C/T]TCTGCTCAAGGCTGGGCGTGGTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612105 MIM: 601728 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLLN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KLLN - killin, p53-regulated DNA replication inhibitor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PTEN - phosphatase and tensin homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000314.6 | Intron | NP_000305.3 | ||||
NM_001304717.2 | Intron | NP_001291646.2 | ||||
NM_001304718.1 | Intron | NP_001291647.1 |