Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTGTTTGCAGGCCTCAGCCAGGT[A/G]CACCTGCAGGGCAATGGGCATAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNU6-6P PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RNU6-6P - RNA, U6 small nuclear 6, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UCMA - upper zone of growth plate and cartilage matrix associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303118.1 | Intron | NP_001290047.1 | ||||
NM_001303119.1 | Intron | NP_001290048.1 | ||||
NM_145314.2 | Intron | NP_660357.2 |