Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCTCTTTATTTGTTAAAAAAGTG[C/G]AAGACGTCCATGCTGTAGAGATTAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
SLC35G1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC35G1 - solute carrier family 35 member G1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134658.1 | 303 | Missense Mutation | CAA,GAA | Q,E 93 | NP_001128130.1 | |
NM_153226.2 | 303 | Missense Mutation | CAA,GAA | Q,E 92 | NP_694958.1 | |
XM_011539371.2 | 303 | Missense Mutation | CAA,GAA | Q,E 76 | XP_011537673.1 | |
XM_011539373.2 | 303 | Missense Mutation | CAA,GAA | Q,E 93 | XP_011537675.1 |