Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTTGGCCTGCATGTGTCTGCCGT[A/G]GAGGCTGCAAAGACAGAGAAAGGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 179555 | ||||||||||||||||||||
Literature Links: |
RSU1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RSU1 - Ras suppressor protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012425.3 | 879 | Missense Mutation | CAC,TAC | H,Y 246 | NP_036557.1 | |
NM_152724.2 | 879 | Missense Mutation | CAC,TAC | H,Y 193 | NP_689937.2 | |
XM_005252552.3 | 879 | Silent Mutation | TCC,TCT | S,S 201 | XP_005252609.1 |