Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGCCATTTATTATATTTGTCTATG[G/T]GGAAAAATTCCCTCAGAATAAATAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 102750 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADK - adenosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001123.3 | Intron | NP_001114.2 | ||||
NM_001202449.1 | Intron | NP_001189378.1 | ||||
NM_001202450.1 | Intron | NP_001189379.1 | ||||
NM_006721.3 | Intron | NP_006712.2 | ||||
XM_017015698.1 | Intron | XP_016871187.1 | ||||
XM_017015699.1 | Intron | XP_016871188.1 | ||||
XM_017015700.1 | Intron | XP_016871189.1 | ||||
XM_017015701.1 | Intron | XP_016871190.1 | ||||
XM_017015702.1 | Intron | XP_016871191.1 | ||||
XM_017015703.1 | Intron | XP_016871192.1 | ||||
XM_017015704.1 | Intron | XP_016871193.1 | ||||
XM_017015705.1 | Intron | XP_016871194.1 | ||||
XM_017015706.1 | Intron | XP_016871195.1 |