Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTCTAGGTTGCTGGATGTCACCC[C/T]CACCTCCAAGTTTGAGGACAACCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600827 | ||||||||||||||||||||
Literature Links: |
PDE6C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PDE6C - phosphodiesterase 6C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006204.3 | 494 | Missense Mutation | CCC,CTC | P,L 119 | NP_006195.3 | |
XM_017016322.1 | 494 | Missense Mutation | CCC,CTC | P,L 22 | XP_016871811.1 |