Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTATGATTCCCTGGATTATGCAAA[A/G]AAAAATGAACCCAAACATAGACTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614258 MIM: 602412 | ||||||||||||||||||||
Literature Links: |
POLR3A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
POLR3A - RNA polymerase III subunit A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPS24 - ribosomal protein S24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001026.4 | 391 | Intron | NP_001017.1 | |||
NM_001142282.1 | 391 | Intron | NP_001135754.1 | |||
NM_001142283.1 | 391 | Intron | NP_001135755.1 | |||
NM_001142284.1 | 391 | Intron | NP_001135756.1 | |||
NM_001142285.1 | 391 | Silent Mutation | AAA,AAG | K,K 83 | NP_001135757.1 | |
NM_033022.3 | 391 | Intron | NP_148982.1 |