Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTGCATTGGCTGGGTATGAATTC[C/G]CTGTGGGCCAAAGGAAAACAGGTAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607318 | ||||||||||||||||||||
Literature Links: |
LOXL4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOXL4 - lysyl oxidase like 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032211.6 | 2518 | Missense Mutation | GCG,GGG | A,G 734 | NP_115587.6 | |
XM_005270216.1 | 2518 | Missense Mutation | GCG,GGG | A,G 735 | XP_005270273.1 | |
XM_006718020.1 | 2518 | Missense Mutation | GCG,GGG | A,G 735 | XP_006718083.1 |
R3HCC1L - R3H domain and coiled-coil containing 1 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |