Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGATGGGTGGGCTCACCAAGTATGG[C/T]ACTGCGTGAAGGAAGTATACCACAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611910 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC16A12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC16A12 - solute carrier family 16 member 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_213606.3 | 1449 | Silent Mutation | GTA,GTG | V,V 420 | NP_998771.3 | |
XM_017016237.1 | 1449 | Silent Mutation | GTA,GTG | V,V 420 | XP_016871726.1 | |
XM_017016238.1 | 1449 | Silent Mutation | GTA,GTG | V,V 420 | XP_016871727.1 | |
XM_017016239.1 | 1449 | Silent Mutation | GTA,GTG | V,V 420 | XP_016871728.1 |