Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGGGCCGCCGGTGCCCTTGCCCG[C/T]CGCGGAGCAGGGTCCTGCGGGGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TBC1D12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TBC1D12 - TBC1 domain family member 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015188.1 | 8 | Missense Mutation | GCC,GTC | A,V 300 | NP_056003.1 | |
XM_006717732.3 | 8 | Missense Mutation | GCC,GTC | A,V 300 | XP_006717795.2 | |
XM_006717733.2 | 8 | UTR 5 | XP_006717796.1 | |||
XM_011539558.2 | 8 | Intron | XP_011537860.1 | |||
XM_017015996.1 | 8 | Missense Mutation | GCC,GTC | A,V 300 | XP_016871485.1 | |
XM_017015997.1 | 8 | Missense Mutation | GCC,GTC | A,V 300 | XP_016871486.1 | |
XM_017015998.1 | 8 | Intron | XP_016871487.1 | |||
XM_017015999.1 | 8 | Intron | XP_016871488.1 |