Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCACCTCACTTCGAGTGACCACA[A/C]TCCGGACCAGGGTGGAGTCATCTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 186360 | ||||||||||||||||||||
Literature Links: |
ANXA7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANXA7 - annexin A7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001156.4 | 1400 | Missense Mutation | NP_001147.1 | |||
NM_001320879.1 | 1400 | Missense Mutation | NP_001307808.1 | |||
NM_001320880.1 | 1400 | Missense Mutation | NP_001307809.1 | |||
NM_004034.3 | 1400 | Missense Mutation | NP_004025.1 | |||
XM_017016162.1 | 1400 | Missense Mutation | XP_016871651.1 | |||
XM_017016163.1 | 1400 | Missense Mutation | XP_016871652.1 |