Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGTGGTTACCCTGCGCCCTTCCT[C/T]TGGGTGGCTGCAGTTCAGCCTCTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604647 | ||||||||||||||||||||
Literature Links: |
CALY PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CALY - calcyon neuron specific vesicular protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321617.1 | 1435 | UTR 5 | NP_001308546.1 | |||
NM_015722.3 | 1435 | Missense Mutation | AAG,GAG | K,E 78 | NP_056537.1 | |
XM_005252685.4 | 1435 | Missense Mutation | AAG,GAG | K,E 78 | XP_005252742.2 | |
XM_011539836.2 | 1435 | Missense Mutation | AAG,GAG | K,E 78 | XP_011538138.1 | |
XM_011539838.2 | 1435 | UTR 5 | XP_011538140.1 |