Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTTTTCTACAATCACCTCTCCAT[A/G]AAGCACCTTTCTTCTTTGACGTAAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610467 | ||||||||||||||||||||
Literature Links: |
HACD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HACD1 - 3-hydroxyacyl-CoA dehydratase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014241.3 | 870 | Missense Mutation | CAT,TAT | H,Y 279 | NP_055056.3 | |
XM_005252641.4 | 870 | Missense Mutation | CAT,TAT | H,Y 243 | XP_005252698.1 |