Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCTCCTTCAGTCACAGTGTTTGA[C/T]CACCATGCCTTTCCCTCAATCTCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601568 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADD3 - adducin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001121.3 | Intron | NP_001112.2 | ||||
NM_001320591.1 | Intron | NP_001307520.1 | ||||
NM_001320592.1 | Intron | NP_001307521.1 | ||||
NM_001320593.1 | Intron | NP_001307522.1 | ||||
NM_001320594.1 | Intron | NP_001307523.1 | ||||
NM_016824.4 | Intron | NP_058432.1 | ||||
NM_019903.4 | Intron | NP_063968.1 |
ADD3-AS1 - ADD3 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |