Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCCCCCATGTCATAGCCAACCAT[A/G]CAGTTCTTGGTGGATGCAGTGAAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPXM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CPXM2 - carboxypeptidase X (M14 family), member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198148.2 | 2035 | Silent Mutation | TGC,TGT | C,C 703 | NP_937791.2 | |
XM_005269528.3 | 2035 | Intron | XP_005269585.1 | |||
XM_011539283.2 | 2035 | Intron | XP_011537585.1 | |||
XM_011539285.2 | 2035 | Silent Mutation | TGC,TGT | C,C 594 | XP_011537587.1 | |
XM_011539286.1 | 2035 | Silent Mutation | TGC,TGT | C,C 476 | XP_011537588.1 | |
XM_017015673.1 | 2035 | Silent Mutation | TGC,TGT | C,C 594 | XP_016871162.1 | |
XM_017015674.1 | 2035 | Silent Mutation | TGC,TGT | C,C 376 | XP_016871163.1 |