Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCGACTTTTTCTTTGTAAGATGTA[A/C]GCCAAAGGATCCGATTTGCATAGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608538 | ||||||||||||||||||||
Literature Links: |
ARID5B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARID5B - AT-rich interaction domain 5B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244638.1 | 596 | Intron | NP_001231567.1 | |||
NM_032199.2 | 596 | Missense Mutation | AAG,ACG | K,T 47 | NP_115575.1 | |
XM_011540262.1 | 596 | Missense Mutation | AAG,ACG | K,T 47 | XP_011538564.1 | |
XM_017016765.1 | 596 | Intron | XP_016872254.1 |