Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCTTGTCGACGTGGGTCCTGGCC[A/G]GCGCCGAGATCACTTGGGACGCGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606285 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SORCS3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SORCS3 - sortilin related VPS10 domain containing receptor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014978.2 | 321 | Missense Mutation | AGC,GGC | S,G 32 | NP_055793.1 | |
XM_011539542.2 | 321 | Intron | XP_011537844.1 |