Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCACACCCGGACCCACTGGTTGCT[C/G]TTGGGCCTTGCTTTGCTCTGCAGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616616 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSGALNACT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CSGALNACT2 - chondroitin sulfate N-acetylgalactosaminyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319654.1 | 424 | Silent Mutation | CTC,CTG | L,L 16 | NP_001306583.1 | |
NM_001319656.1 | 424 | Silent Mutation | CTC,CTG | L,L 16 | NP_001306585.1 | |
NM_018590.4 | 424 | Silent Mutation | CTC,CTG | L,L 16 | NP_061060.3 | |
XM_011539938.1 | 424 | Silent Mutation | CTC,CTG | L,L 16 | XP_011538240.1 | |
XM_017016388.1 | 424 | Silent Mutation | CTC,CTG | L,L 16 | XP_016871877.1 | |
XM_017016389.1 | 424 | Silent Mutation | CTC,CTG | L,L 16 | XP_016871878.1 | |
XM_017016390.1 | 424 | Silent Mutation | CTC,CTG | L,L 16 | XP_016871879.1 | |
XM_017016391.1 | 424 | Silent Mutation | CTC,CTG | L,L 16 | XP_016871880.1 |