Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTTCTCATGAAGATATTTTAAAC[C/T]TCCATGCTCTGAGAAATCTGAGACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 138250 | ||||||||||||||||||||
Literature Links: |
ALDH18A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ALDH18A1 - aldehyde dehydrogenase 18 family member A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017423.1 | 2597 | Missense Mutation | AGT,GGT | S,G 777 | NP_001017423.1 | |
NM_001323412.1 | 2597 | Missense Mutation | AGT,GGT | S,G 668 | NP_001310341.1 | |
NM_001323413.1 | 2597 | Missense Mutation | AGT,GGT | S,G 779 | NP_001310342.1 | |
NM_001323414.1 | 2597 | Missense Mutation | AGT,GGT | S,G 779 | NP_001310343.1 | |
NM_001323415.1 | 2597 | Missense Mutation | AGT,GGT | S,G 777 | NP_001310344.1 | |
NM_001323416.1 | 2597 | Missense Mutation | AGT,GGT | S,G 668 | NP_001310345.1 | |
NM_001323417.1 | 2597 | Missense Mutation | AGT,GGT | S,G 744 | NP_001310346.1 | |
NM_001323418.1 | 2597 | Missense Mutation | AGT,GGT | S,G 666 | NP_001310347.1 | |
NM_001323419.1 | 2597 | Missense Mutation | AGT,GGT | S,G 567 | NP_001310348.1 | |
NM_002860.3 | 2597 | Missense Mutation | AGT,GGT | S,G 779 | NP_002851.2 |