Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTTGTATGCCATAGTATTGTGCA[A/G]GTAAGTCTTTGAAGTAGGTGTTTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TMEM254 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMEM254 - transmembrane protein 254 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270367.1 | 400 | Missense Mutation | AAG,AGG | K,R 108 | NP_001257296.1 | |
NM_001270368.1 | 400 | Missense Mutation | AAG,AGG | K,R 108 | NP_001257297.1 | |
NM_001270369.1 | 400 | Missense Mutation | AAA,AGA | K,R 108 | NP_001257298.1 | |
NM_001270370.1 | 400 | Missense Mutation | AAG,AGG | K,R 84 | NP_001257299.1 | |
NM_001270371.1 | 400 | UTR 3 | NP_001257300.1 | |||
NM_001270372.1 | 400 | Missense Mutation | AAG,AGG | K,R 84 | NP_001257301.1 | |
NM_001270373.1 | 400 | Missense Mutation | AAA,AGA | K,R 84 | NP_001257302.1 | |
NM_001270374.1 | 400 | UTR 3 | NP_001257303.1 | |||
NM_025125.3 | 400 | Missense Mutation | AAG,AGG | K,R 84 | NP_079401.2 | |
XM_006717978.2 | 400 | Missense Mutation | AAG,AGG | K,R 93 | XP_006718041.1 | |
XM_011540192.2 | 400 | Missense Mutation | AAG,AGG | K,R 108 | XP_011538494.1 | |
XM_011540193.2 | 400 | UTR 3 | XP_011538495.1 | |||
XM_017016679.1 | 400 | Missense Mutation | AAG,AGG | K,R 108 | XP_016872168.1 | |
XM_017016680.1 | 400 | Missense Mutation | AAG,AGG | K,R 93 | XP_016872169.1 |
TMEM254-AS1 - TMEM254 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |