Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTGACGTGGGGTTCTCCGATTTG[C/T]TTTCTTGGCTGGACGGAGGCTTGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603451 | ||||||||||||||||||||
Literature Links: |
LDB1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LDB1 - LIM domain binding 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001113407.2 | 853 | Missense Mutation | AAC,AGC | N,S 400 | NP_001106878.1 | |
NM_001321612.1 | 853 | Missense Mutation | AAC,AGC | N,S 398 | NP_001308541.1 | |
NM_003893.4 | 853 | Missense Mutation | AAC,AGC | N,S 364 | NP_003884.1 | |
XM_017016867.1 | 853 | Intron | XP_016872356.1 | |||
XM_017016868.1 | 853 | Intron | XP_016872357.1 | |||
XM_017016869.1 | 853 | Missense Mutation | AAC,AGC | N,S 231 | XP_016872358.1 |