Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAATTAGGGGCCTGTCGAGCGAT[A/G]TTGGAAGAAGAAACAGATCAAACCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609672 | ||||||||||||||||||||
Literature Links: |
EXOC6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EXOC6 - exocyst complex component 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013848.3 | 204 | Missense Mutation | ATA,ATG | I,M 1 | NP_001013870.1 | |
NM_001319194.1 | 204 | Missense Mutation | ATA,ATG | I,M 22 | NP_001306123.1 | |
NM_001319195.1 | 204 | Intron | NP_001306124.1 | |||
NM_001319200.1 | 204 | Intron | NP_001306129.1 | |||
NM_019053.5 | 204 | Intron | NP_061926.3 | |||
XM_017016344.1 | 204 | Missense Mutation | ATA,ATG | I,M 22 | XP_016871833.1 | |
XM_017016345.1 | 204 | Missense Mutation | ATA,ATG | I,M 1 | XP_016871834.1 | |
XM_017016346.1 | 204 | Missense Mutation | ATA,ATG | I,M 1 | XP_016871835.1 | |
XM_017016347.1 | 204 | Intron | XP_016871836.1 | |||
XM_017016348.1 | 204 | Missense Mutation | ATA,ATG | I,M 22 | XP_016871837.1 |