Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGAGCAGTTTGAAAGGAAACTAT[A/C]CTCACCAAAAGAATCTAAACCCAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610348 | ||||||||||||||||||||
Literature Links: |
SLF2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLF2 - SMC5-SMC6 complex localization factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136123.1 | 823 | Missense Mutation | TAC,TCC | Y,S 94 | NP_001129595.1 | |
NM_001243770.1 | 823 | Intron | NP_001230699.1 | |||
NM_018121.3 | 823 | Missense Mutation | TAC,TCC | Y,S 94 | NP_060591.3 | |
XM_005269965.2 | 823 | Missense Mutation | TAC,TCC | Y,S 94 | XP_005270022.1 | |
XM_011539944.2 | 823 | Missense Mutation | TAC,TCC | Y,S 30 | XP_011538246.1 |