Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGCCTTCATCATCCTGCGTAAGC[A/G]GCCACTCATCTTTATTCACTGGTAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611815 MIM: 602669 | ||||||||||||||||||||
Literature Links: |
ELOVL3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ELOVL3 - ELOVL fatty acid elongase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152310.2 | 1677 | Missense Mutation | CAG,CGG | Q,R 139 | NP_689523.1 | |
XM_011540245.2 | 1677 | Missense Mutation | CAG,CGG | Q,R 139 | XP_011538547.1 |
PITX3 - paired like homeodomain 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |