Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTGAGCGAGGCTGTGGTGAGCAGC[A/G]TCAACACTGTGGCCACCAAGACCGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608925 MIM: 602998 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADIRF PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADIRF - adipogenesis regulatory factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MMRN2 - multimerin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024756.2 | 387 | Intron | NP_079032.2 | |||
XM_005270153.1 | 387 | Intron | XP_005270210.1 | |||
XM_006717970.3 | 387 | Intron | XP_006718033.1 |
SNCG - synuclein gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003087.2 | 387 | Missense Mutation | ATC,GTC | I,V 74 | NP_003078.2 | |
XM_011540105.2 | 387 | Missense Mutation | CAT,CGT | H,R 91 | XP_011538407.1 |