Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAGCAGGACACTCTCTCTCATGGA[A/C]TGATGAAAAACAACTCAAGGAACAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607194 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C10orf67 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C10orf67 - chromosome 10 open reading frame 67 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PTF1A - pancreas specific transcription factor, 1a | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178161.2 | 851 | Missense Mutation | AAT,ACT | N,T 284 | NP_835455.1 |