Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCTGGTTCTTTGGAGGGATCACC[A/G]CCTCGTTCCAGGTACAGATTATTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602140 MIM: 610258 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NDUFB8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NDUFB8 - NADH:ubiquinone oxidoreductase subunit B8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284367.1 | 795 | UTR 3 | NP_001271296.1 | |||
NM_001284368.1 | 795 | Intron | NP_001271297.1 | |||
NM_005004.3 | 795 | Silent Mutation | GGC,GGT | G,G 171 | NP_004995.1 |
SEC31B - SEC31 homolog B, COPII coat complex component | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |