Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTAAAGTTCATTTCCATTCACACA[C/T]TTTTGCTGCTGAAGCCTCGTTCTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609389 MIM: 610909 | ||||||||||||||||||||
Literature Links: |
INPP5F PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
INPP5F - inositol polyphosphate-5-phosphatase F | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MCMBP - minichromosome maintenance complex binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256378.1 | 2045 | Silent Mutation | AAA,AAG | K,K 633 | NP_001243307.1 | |
NM_001256379.1 | 2045 | Silent Mutation | AAA,AAG | K,K 460 | NP_001243308.1 | |
NM_024834.3 | 2045 | Silent Mutation | AAA,AAG | K,K 635 | NP_079110.1 | |
XM_005270157.4 | 2045 | Silent Mutation | AAA,AAG | K,K 580 | XP_005270214.1 | |
XM_011540170.1 | 2045 | Silent Mutation | AAA,AAG | K,K 565 | XP_011538472.1 | |
XM_017016663.1 | 2045 | Silent Mutation | AAA,AAG | K,K 578 | XP_016872152.1 | |
XM_017016664.1 | 2045 | Silent Mutation | AAA,AAG | K,K 563 | XP_016872153.1 |